Assay Details
| Gene Symbol: ACP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722062.3 | XP_002722108.1 | 284 | 69 | ||
| XM_017338010.1 | XP_017193499.1 | 288 | 69 | |||
| XM_017338011.1 | XP_017193500.1 | 284 | 69 | |||
| XM_017338013.1 | XP_017193502.1 | 222 | 69 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017338013.1 transcripts.
Set Membership: |
|

ACP3
Genomic Map