Assay Details
| Gene Symbol: ACP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722062.3 | XP_002722108.1 | 522 | 71 | ||
| XM_017338010.1 | XP_017193499.1 | 526 | 71 | |||
| XM_017338011.1 | XP_017193500.1 | 522 | 71 | |||
| XM_017338012.1 | XP_017193501.1 | 441 | 71 | |||
| XM_017338013.1 | XP_017193502.1 | 460 | 71 | |||
More Information
Set Membership: |
|

ACP3
Genomic Map