Assay Details
| Gene Symbol: ACP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722062.3 | XP_002722108.1 | 854 | 98 | ||
| XM_017338010.1 | XP_017193499.1 | 858 | 98 | |||
| XM_017338011.1 | XP_017193500.1 | 854 | 98 | |||
| XM_017338012.1 | XP_017193501.1 | 773 | 98 | |||
| XM_017338013.1 | XP_017193502.1 | 792 | 98 | |||
More Information
Set Membership: |
|

ACP3
Genomic Map