Assay Details
Gene Symbol: LOC100343106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008251255.2 | XP_008249477.1 | 136 | 65 | ||
XM_017339105.1 | XP_017194594.1 | 251 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008251255.2 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
