Assay Details
| Gene Symbol: C9H18orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344152.1 | XP_017199641.1 | 1 - 2 | 170 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017344152.1 transcripts.
Set Membership: |
|

C9H18orf25
Genomic Map