Assay Details
| Gene Symbol: LOC100354908 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723116.3 | XP_002723162.1 | 1867 | 80 | ||
| XM_017339128.1 | XP_017194617.1 | 1583 | 80 | |||
More Information
Set Membership: |
|

LOC100354908
Genomic Map