Assay Details
| Gene Symbol: LOC100342021 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008253649.2 | XP_008251871.1 | 8 - 9 | 1335 | 74 | |
| XM_008253656.2 | XP_008251878.1 | 8 - 9 | 1425 | 74 | ||
| XM_017339820.1 | XP_017195309.1 | 8 - 9 | 1241 | 74 | ||
More Information
Set Membership: |
|

LOC100342021
Genomic Map