Assay Details
| Gene Symbol: LOC100338055 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712677.3 | XP_002712723.2 | 1 - 2 | 705 | 68 | |
| XM_017343275.1 | XP_017198764.1 | 1 - 2 | 713 | 68 | ||
| XM_017343276.1 | XP_017198765.1 | 1 - 2 | 708 | 68 | ||
More Information
Set Membership: |
|

LOC100338055
Genomic Map