Assay Details
| Gene Symbol: LOC100338055 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712677.3 | XP_002712723.2 | 2 - 3 | 894 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100338055| Gene Symbol: LOC100338055 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712677.3 | XP_002712723.2 | 2 - 3 | 894 | 85 | |
Set Membership: |
|