Assay Details
| Gene Symbol: FHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716904.3 | XP_002716950.1 | 7 - 8 | 1128 | 54 | |
| XM_008267310.2 | XP_008265532.1 | 7 - 8 | 1164 | 54 | ||
| XM_008267312.2 | XP_008265534.1 | 6 - 7 | 576 | 54 | ||
More Information
Set Membership: |
|

FHDC1
Genomic Map