Assay Details
| Gene Symbol: LRRC46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719124.3 | XP_002719170.2 | 3 - 4 | 450 | 61 | |
| XM_017349173.1 | XP_017204662.1 | 3 - 4 | 452 | 61 | ||
More Information
Set Membership: |
|

LRRC46
Genomic Map