Assay Details
| Gene Symbol: GNG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721000.3 | XP_002721046.1 | 294 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002721000.3 transcripts.
Set Membership: |
|

GNG3
Genomic Map