Assay Details
| Gene Symbol: EDIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261919.2 | XP_008260141.1 | 9 - 10 | 1726 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EDIL3| Gene Symbol: EDIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261919.2 | XP_008260141.1 | 9 - 10 | 1726 | 60 | |
Set Membership: |
|