Assay Details
| Gene Symbol: PDCD2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711505.3 | XP_002711551.1 | 1 - 2 | 138 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PDCD2L| Gene Symbol: PDCD2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711505.3 | XP_002711551.1 | 1 - 2 | 138 | 78 | |
Set Membership: |
|