Assay Details
| Gene Symbol: MCOLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008264961.2 | XP_008263183.1 | 6 - 7 | 893 | 59 | |
| XM_008264962.2 | XP_008263184.1 | 6 - 7 | 772 | 59 | ||
More Information
Set Membership: |
|

MCOLN2
Genomic Map