Assay Details
| Gene Symbol: PTPDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002708293.3 | XP_002708339.2 | 2 - 3 | 511 | 65 | |
| XM_008257290.2 | XP_008255512.1 | 3 - 4 | 346 | 65 | ||
| XM_008257292.2 | XP_008255514.1 | 3 - 4 | 560 | 65 | ||
| XM_017342121.1 | XP_017197610.1 | 2 - 3 | 525 | 65 | ||
| XM_017342122.1 | XP_017197611.1 | 2 - 3 | 523 | 65 | ||
| XM_017342124.1 | XP_017197613.1 | 2 - 3 | 523 | 65 | ||
| XM_017342127.1 | XP_017197616.1 | 2 - 3 | 522 | 65 | ||
| XM_017342129.1 | XP_017197618.1 | 2 - 3 | 522 | 65 | ||
| XM_017342132.1 | XP_017197621.1 | 3 - 4 | 368 | 65 | ||
More Information
Set Membership: |
|

PTPDC1
Genomic Map