Assay Details
| Gene Symbol: LAX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268554.2 | XP_008266776.1 | 3 - 4 | 2162 | 86 | |
| XM_008268555.2 | XP_008266777.1 | 4 - 5 | 530 | 86 | ||
| XM_017347842.1 | XP_017203331.1 | 4 - 5 | 502 | 86 | ||
| XM_017347843.1 | XP_017203332.1 | 3 - 4 | 388 | 86 | ||
More Information
Set Membership: |
|

LAX1
Genomic Map