Assay Details
| Gene Symbol: CDC20B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262208.2 | XP_008260430.1 | 3 - 4 | 491 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDC20B| Gene Symbol: CDC20B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262208.2 | XP_008260430.1 | 3 - 4 | 491 | 58 | |
Set Membership: |
|