Assay Details
| Gene Symbol: KIF2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715148.3 | XP_002715194.1 | 19 - 20 | 2078 | 65 | |
| XM_008265356.2 | XP_008263578.1 | 18 - 19 | 1955 | 65 | ||
| XM_008265357.2 | XP_008263579.1 | 18 - 19 | 2155 | 65 | ||
More Information
Set Membership: |
|

KIF2C
Genomic Map