Assay Details
| Gene Symbol: ZNF691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008265436.2 | XP_008263658.1 | 2 | 1253 | 74 | |
| XM_017346392.1 | XP_017201881.1 | 2 | 1433 | 74 | ||
More Information
Set Membership: |
|

ZNF691
Genomic Map