Assay Details
| Gene Symbol: SKAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271352.2 | XP_008269574.1 | 8 - 9 | 690 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SKAP1| Gene Symbol: SKAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008271352.2 | XP_008269574.1 | 8 - 9 | 690 | 76 | |
Set Membership: |
|