Assay Details
Gene Symbol: SHLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017348685.1 | XP_017204174.1 | 2 - 3 | 2403 | 78 | |
XM_017348686.1 | XP_017204175.1 | 2 - 3 | 2406 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
