Assay Details
| Gene Symbol: GFAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349244.1 | XP_017204733.1 | 9 - 10 | 1404 | 59 | |
| XM_017349245.1 | XP_017204734.1 | 8 - 9 | 1281 | 59 | ||
More Information
Set Membership: |
|

GFAP
Genomic Map