Assay Details
| Gene Symbol: PRICKLE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713291.3 | XP_002713337.1 | 8 - 9 | 1917 | 95 | |
| XM_008260882.2 | XP_008259104.1 | 8 - 9 | 2025 | 95 | ||
| XM_008260883.2 | XP_008259105.1 | 7 - 8 | 2453 | 95 | ||
| XM_008260884.2 | XP_008259106.1 | 7 - 8 | 1978 | 95 | ||
More Information
Set Membership: |
|

PRICKLE2
Genomic Map