Assay Details
| Gene Symbol: NEFH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008274638.2 | XP_008272860.1 | 311 | 98 | ||
| XM_008274639.2 | XP_008272861.1 | 311 | 98 | |||
| XM_008274642.2 | XP_008272864.1 | 312 | 98 | |||
| XM_008274644.2 | XP_008272866.1 | 311 | 98 | |||
| XM_008274645.2 | XP_008272867.1 | 311 | 98 | |||
| XM_008274648.2 | XP_008272870.1 | 311 | 98 | |||
| XM_008274649.2 | XP_008272871.1 | 312 | 98 | |||
| XM_008274650.2 | XP_008272872.1 | 312 | 98 | |||
| XM_008274656.2 | XP_008272878.1 | 311 | 98 | |||
| XM_008274657.2 | XP_008272879.1 | 312 | 98 | |||
| XM_008274660.2 | XP_008272882.1 | 312 | 98 | |||
| XM_008274663.2 | XP_008272885.1 | 312 | 98 | |||
| XM_008274665.2 | XP_008272887.1 | 311 | 98 | |||
| XM_008274668.2 | XP_008272890.1 | 311 | 98 | |||
| XM_008274676.2 | XP_008272898.1 | 313 | 98 | |||
| XM_017350679.1 | XP_017206168.1 | 311 | 98 | |||
| XM_017350680.1 | XP_017206169.1 | 311 | 98 | |||
More Information
Set Membership: |
|

NEFH
Genomic Map