Assay Details
| Gene Symbol: CALY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718769.3 | XP_002718815.1 | 3 - 4 | 377 | 59 | |
| XM_008270646.1 | XP_008268868.1 | 3 - 4 | 378 | 59 | ||
| XM_008270647.2 | XP_008268869.1 | 3 - 4 | 413 | 59 | ||
More Information
Set Membership: |
|

CALY
Genomic Map