Assay Details
| Gene Symbol: MRPL22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710180.3 | XP_002710226.1 | 5 - 6 | 415 | 82 | |
| XM_008255300.2 | XP_008253522.1 | 6 - 7 | 618 | 82 | ||
| XM_008255301.2 | XP_008253523.1 | 6 - 7 | 448 | 82 | ||
| XM_008255303.2 | XP_008253525.1 | 5 - 6 | 361 | 82 | ||
| XM_008255304.2 | XP_008253526.1 | 5 - 6 | 411 | 82 | ||
| XM_008255305.2 | XP_008253527.1 | 6 - 7 | 556 | 82 | ||
| XM_008255306.2 | XP_008253528.1 | 4 - 5 | 350 | 82 | ||
| XM_017341212.1 | XP_017196701.1 | 3 - 4 | 191 | 82 | ||
| XM_017341214.1 | XP_017196703.1 | 6 - 7 | 581 | 82 | ||
More Information
Set Membership: |
|

MRPL22
Genomic Map