Assay Details
| Gene Symbol: LRRC34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008266460.2 | XP_008264682.1 | 7 - 8 | 1059 | 64 | |
| XM_008266462.2 | XP_008264684.1 | 6 - 7 | 976 | 64 | ||
| XM_017346909.1 | XP_017202398.1 | 7 - 8 | 1145 | 64 | ||
More Information
Set Membership: |
|

LRRC34
Genomic Map