Assay Details
| Gene Symbol: RAB6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716590.3 | XP_002716636.2 | 7 - 8 | 571 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB6B| Gene Symbol: RAB6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716590.3 | XP_002716636.2 | 7 - 8 | 571 | 63 | |
Set Membership: |
|