Assay Details
| Gene Symbol: SEPTIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721381.3 | XP_002721427.1 | 1178 | 71 | ||
| XM_008275153.2 | XP_008273375.1 | 2724 | 71 | |||
| XM_008275154.2 | XP_008273376.1 | 2722 | 71 | |||
| XM_008275156.2 | XP_008273378.1 | 1176 | 71 | |||
More Information
Set Membership: |
|

SEPTIN3
Genomic Map