Assay Details
| Gene Symbol: MARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712268.3 | XP_002712314.1 | 1 | 2415 | 126 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MARS2| Gene Symbol: MARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712268.3 | XP_002712314.1 | 1 | 2415 | 126 | |
Set Membership: |
|