Assay Details
| Gene Symbol: ERICH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008258724.2 | XP_008256946.1 | 5 - 6 | 634 | 64 | |
| XM_008258725.2 | XP_008256947.1 | 2 - 3 | 269 | 64 | ||
More Information
Set Membership: |
|

ERICH2
Genomic Map