Assay Details
| Gene Symbol: HPS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008275122.2 | XP_008273344.1 | 2083 | 62 | ||
| XM_008275123.2 | XP_008273345.1 | 1929 | 62 | |||
| XM_008275124.2 | XP_008273346.1 | 2062 | 62 | |||
| XM_017350900.1 | XP_017206389.1 | 2083 | 62 | |||
| XM_017350901.1 | XP_017206390.1 | 2083 | 62 | |||
More Information
Set Membership: |
|

HPS5
Genomic Map