Assay Details
| Gene Symbol: TBCCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716459.3 | XP_002716505.1 | 4 - 5 | 985 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TBCCD1| Gene Symbol: TBCCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716459.3 | XP_002716505.1 | 4 - 5 | 985 | 68 | |
Set Membership: |
|