Assay Details
| Gene Symbol: RIPOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017342299.1 | XP_017197788.1 | 12 - 13 | 1813 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RIPOR1| Gene Symbol: RIPOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017342299.1 | XP_017197788.1 | 12 - 13 | 1813 | 76 | |
Set Membership: |
|