Assay Details
| Gene Symbol: IRF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008253698.2 | XP_008251920.1 | 805 | 75 | ||
| XM_008253699.2 | XP_008251921.1 | 802 | 75 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IRF7| Gene Symbol: IRF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008253698.2 | XP_008251920.1 | 805 | 75 | ||
| XM_008253699.2 | XP_008251921.1 | 802 | 75 | |||
Set Membership: |
|