Assay Details
| Gene Symbol: NCBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716527.3 | XP_002716573.1 | 3 - 4 | 512 | 72 | |
| XM_008266708.2 | XP_008264930.1 | 5 - 6 | 781 | 72 | ||
| XM_008266709.2 | XP_008264931.1 | 5 - 6 | 778 | 72 | ||
| XM_008266710.2 | XP_008264932.1 | 3 - 4 | 456 | 72 | ||
| XM_017347011.1 | XP_017202500.1 | 4 - 5 | 1426 | 72 | ||
More Information
Set Membership: |
|

NCBP2
Genomic Map