Assay Details
| Gene Symbol: PQBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008272594.2 | XP_008270816.1 | 4 - 5 | 368 | 64 | |
| XM_008272595.2 | XP_008270817.1 | 3 - 4 | 323 | 64 | ||
| XM_008272596.2 | XP_008270818.1 | 3 - 4 | 291 | 64 | ||
More Information
Set Membership: |
|

PQBP1
Genomic Map