Assay Details
| Gene Symbol: FN3K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723048.3 | XP_002723094.1 | 296 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FN3K| Gene Symbol: FN3K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723048.3 | XP_002723094.1 | 296 | 72 | ||
Set Membership: |
|