Assay Details
| Gene Symbol: KIF3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008256144.2 | XP_008254366.1 | 6 - 7 | 2043 | 64 | |
| XM_017341699.1 | XP_017197188.1 | 5 - 6 | 1860 | 64 | ||
More Information
Set Membership: |
|

KIF3B
Genomic Map