Assay Details
| Gene Symbol: ABHD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713272.3 | XP_002713318.2 | 6 - 7 | 800 | 64 | |
| XM_008260838.2 | XP_008259060.1 | 6 - 7 | 824 | 64 | ||
| XM_008260839.2 | XP_008259061.1 | 6 - 7 | 816 | 64 | ||
More Information
Set Membership: |
|

ABHD6
Genomic Map