Assay Details
| Gene Symbol: NR4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261574.2 | XP_008259796.1 | 6 - 7 | 2115 | 65 | |
| XM_008261684.2 | XP_008259906.1 | 5 - 6 | 2043 | 65 | ||
| XM_017349404.1 | XP_017204893.1 | 4 - 5 | 1858 | 65 | ||
| XM_017349418.1 | XP_017204907.1 | 5 - 6 | 2146 | 65 | ||
More Information
Set Membership: |
|

NR4A3
Genomic Map