Assay Details
| Gene Symbol: CAPZA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715766.3 | XP_002715812.1 | 6 - 7 | 885 | 75 | |
| XM_017345978.1 | XP_017201467.1 | 6 - 7 | 888 | 75 | ||
More Information
Set Membership: |
|

CAPZA1
Genomic Map