Assay Details
| Gene Symbol: RPL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710706.2 | XP_002710752.1 | 1 - 2 | 96 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002710706.2 transcripts.
Set Membership: |
|

RPL30
Genomic Map