Assay Details
| Gene Symbol: LOC100350045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720955.3 | XP_002721001.1 | 178 | 125 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002720955.3 transcripts.
Set Membership: |
|

LOC100350045
Genomic Map