Assay Details
| Gene Symbol: RPL37A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712466.3 | XP_002712512.1 | 1 - 2 | 115 | 102 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002712466.3 transcripts.
Set Membership: |
|

RPL37A
Genomic Map