Assay Details
| Gene Symbol: LOC103351326 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008269871.1 | XP_008268093.1 | 1 - 2 | 87 | 74 | |
| XM_017348681.1 | XP_017204170.1 | 1 - 2 | 87 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008269871.1, XM_017348681.1 transcripts.
Set Membership: |
|

LOC103351326
Genomic Map