Assay Details
| Gene Symbol: ADIPOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712774.3 | XP_002712820.1 | 4 | 601 | 64 | |
| XM_017343434.1 | XP_017198923.1 | 4 | 638 | 64 | ||
| XM_017343435.1 | XP_017198924.1 | 4 | 694 | 64 | ||
| XM_017343436.1 | XP_017198925.1 | 5 | 786 | 64 | ||
| GenBank mRNA | AM886135.1 | - | 2 | 124 | 64 | |
More Information
Set Membership: |
|

ADIPOR2
Genomic Map