Assay Details
| Gene Symbol: SS18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713461.3 | XP_002713507.1 | 10 | 1430 | 97 | |
| XM_008261156.2 | XP_008259378.2 | 11 | 1465 | 97 | ||
| XM_008261157.2 | XP_008259379.2 | 10 | 1318 | 97 | ||
| XM_008261161.2 | XP_008259383.1 | 10 | 1300 | 97 | ||
| XM_008261162.2 | XP_008259384.1 | 9 | 1402 | 97 | ||
| XM_008261164.2 | XP_008259386.1 | 11 | 1523 | 97 | ||
| XM_017344091.1 | XP_017199580.1 | 10 | 1406 | 97 | ||
| XM_017344092.1 | XP_017199581.1 | 9 | 1206 | 97 | ||
More Information
Set Membership: |
|

SS18
Genomic Map