Assay Details
| Gene Symbol: CRABP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715374.3 | XP_002715420.1 | 1 | 108 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002715374.3 transcripts.
Set Membership: |
|

CRABP2
Genomic Map